missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Thin Wall 0.2 mL PCR Tubes with Attached Caps

Manufactured without lubricants or chemical additives that can contaminate reagents and samples.Thin Wall 0.2mL PCR Tubes with Attached Caps are autoclavable and RNase-, DNase-, and DNA-free. Thin, uniform walls ensure consistent, precise thermal transfer.

66.00 € - 71.45 €

Specifications

Certifications/Compliance RNase, DNase and DNA free
Volume (Metric) 0.2 mL
Quantity 1000 Pack
For Use With (Equipment) Thermal cyclers for excellent reproducible results in most PCR applications
Sterility Non-sterile
View More Specs

Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10693443
View Documents
BRAND™
781301
Rose
66.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
10571723
View Documents
BRAND™
781302
Yellow
67.20 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
10581723
View Documents
BRAND™
781303
Green
67.20 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
10673253
View Documents
BRAND™
781304
Blue
67.20 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
10161391
View Documents
BRAND™
781305
Clear
71.45 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Manufactured without lubricants or chemical additives that can contaminate reagents and samples
  • Thin, uniform walls ensure consistent, precise thermal transfer
  • Autoclavable
  • RNase-, DNase-, and DNA-free
  • Compatible with most thermal cyclers
  • Attached flat caps that form a tight seal to help prevent sample evaporation but open easily

Specifications

Specifications

RNase, DNase and DNA free
1000 Pack
Non-sterile
Flat Cap
Yes
Tube
Thin Wall PCR Tube
0.2 mL
Thermal cyclers for excellent reproducible results in most PCR applications
Polypropylene
Autoclavable
Single-use
0.2 mL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Thin Wall 0.2 mL PCR Tubes with Attached Caps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.