Learn More
VACUUBRAND™ Pump Oil
Replacement oil for BrandTech Vacuubrand Rotary Vane and Chemistry-Hybrid Vacuum Pumps
39.80 € - 171.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10633952
|
VACUUBRAND™
20687010 |
1 L |
39.80 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11854000
|
VACUUBRAND™
20687011 |
5 L |
171.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rotary vane pumps are used whenever it is necessary to have a process vacuum of up to 10-3 mbar. VACUUBRAND rotary vane pumps are high-performance, yet compact, and can be equipped with an extensive line of VACUUBRAND accessories. They have an innovative lubrication system with a built-in oil pump and have a large oil volume. This extends oil change and service intervals and protects the pump at start-up. The effective gas ballast feature, with its high-flow gas ballast, provides high vapor pumping capability for water and solvents. VACUUBRAND rotary vane pumps´ volume flow rate is specified at atmospheric pressure, as is customary with PNEUROP™. For process effiency, however, the high volume flow rate of VACUUBRAND pumps under process conditions, as well as a consistently high volume flow rate over a wide pressure range, is the key to your satisfaction in real-world application. After switch-off the aggregate is vacuum-sealed to protect your application from undesired venting and oil back flow.- Constriction-free vapor pathway allows high volume flow rates, even when close to the ultimate vacuum
- high tolerance for water and solvent vapors, because of the high-volume gas ballast
- quiet running and excellent ultimate vacuum, even with gas ballast
- active corrosion protection: Oil cycle closes vacuum-tight against the intake of corrosive gases and oil impurities into the reservoir when shut down
- new design lubrication circuit, and large usable oil volume provides extended oil change and service intervals
- compact design, low weight and easy service with telescopic design
Specifications
Rotary vane vacuum pump | |
<1 x 10-3 mbar |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
VACUUBRAND™ Pump Oil