Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Beaker Positioning Covers
For use with Branson ultrasonic cleaning baths
Brand:  Branson Ultrasonics™ 100-032-518
 View more versions of this product
Product Code. 15308634
  • 51.80 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with Branson ultrasonic cleaning baths
  • Helps positon beakers securely in bath
Specifications
Specifications
Beaker Positioning Cover
Beaker in Ultrasonic Benchtop Cleaning Bath, 2 x 600 mL, Fits Model 2800
Helps position beakers securely in bath