missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaning Bath Accessory, Beaker Positioning Covers
For use with Branson ultrasonic cleaning baths
46.45 € - 188.00 €
Specifications
Type | Beaker Positioning Cover |
---|---|
For Use With (Application) | Helps position beakers securely in bath |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15328634
|
Branson Ultrasonics™
100-032-528 |
Beaker in Ultrasonic Benchtop Cleaning Bath, 3 x 250 mL, Fits Model 3800 |
69.00 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
15398624
|
Branson Ultrasonics™
100-032-527 |
Beaker in Ultrasonic Benchtop Cleaning Bath, 2 x 250 mL, Fits Model 2800 |
51.80 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
15338634
|
Branson Ultrasonics™
100-032-520 |
Beaker in Ultrasonic Benchtop Cleaning Bath, 2 x 600 mL, Fits Model 3800 |
54.60 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
15308634
|
Branson Ultrasonics™
100-032-518 |
Beaker in Ultrasonic Benchtop Cleaning Bath, 2 x 600 mL, Fits Model 2800 |
51.80 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
15388634
|
Branson Ultrasonics™
100-032-511 |
Beaker in Ultrasonic Benchtop Cleaning Bath, 4 x 600 mL, Fits Model 5800 |
79.60 €
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
13413649
|
Branson Ultrasonics™
CPN-246-014 |
Beaker in Ultrasonic Benchtop Cleaning Bath, 6 x 600 mL, Fits Model 8800 |
188.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15636458
|
Branson Ultrasonics™
100-032-516 |
Beaker in Ultrasonic Benchtop Cleaning Bath, 1 x 600 mL, Fits Model 1800 |
46.45 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Description
For use with Branson ultrasonic cleaning baths
- Helps positon beakers securely in bath
Specifications
Beaker Positioning Cover | |
Helps position beakers securely in bath |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title