Learn More
Brepocitinib P-Tosylate, MedChemExpress
MedChemExpress Brepocitinib (PF-06700841) P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. Brepocitinib P-Tosylate also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively.
Brand:  Medchem Express HY-112708A
Encompass_Preferred
 View more versions of this product
Product Code. 16420977
  • 743.85 € / 50mg
Explore more special offers
Description
Description
Brepocitinib (PF-06700841) P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. Brepocitinib P-Tosylate also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively.
Specifications
Specifications
Brepocitinib P-Tosylate
White
C25H29F2N7O4S
PF-06700841 P-Tosylate
O=C([C@H]1C(F)(F)C1)N2C3CN(C4=NC(NC5=CN(C)N=C5)=NC=C4)CC2CC3.O=S(C6=CC=C(C)C=C6)(O)=O
561.6
Research
COVID-19-immunoregulation
2140301-96-6
Research
50 mg
DMSO : 62.5 mg/mL (111.29 mM; Need ultrasonic)
561.6
99.69%
Solid