Learn More
Brepocitinib P-Tosylate, MedChemExpress
MedChemExpress Brepocitinib (PF-06700841) P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. Brepocitinib P-Tosylate also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively.
112.05 € - 1115.10 €
Specifications
Chemical Name or Material | Brepocitinib P-Tosylate |
---|---|
CAS | 2140301-96-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C25H29F2N7O4S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16485516
|
Medchem Express
HY-112708A |
100 mg |
1115.10 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16477397
|
Medchem Express
HY-112708A |
5 mg |
112.05 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16424688
|
Medchem Express
HY-112708A |
10 mg |
180.90 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16420977
|
Medchem Express
HY-112708A |
50 mg |
697.95 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Brepocitinib (PF-06700841) P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. Brepocitinib P-Tosylate also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively.Specifications
Brepocitinib P-Tosylate | |
White | |
C25H29F2N7O4S | |
DMSO : 62.5 mg/mL (111.29 mM; Need ultrasonic) | |
561.6 | |
99.69% | |
Solid |
2140301-96-6 | |
Research | |
PF-06700841 P-Tosylate | |
O=C([C@H]1C(F)(F)C1)N2C3CN(C4=NC(NC5=CN(C)N=C5)=NC=C4)CC2CC3.O=S(C6=CC=C(C)C=C6)(O)=O | |
561.6 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.