Learn More
Leica Microsystems DM500 Brightfield Microscope
Ideal tool to make teaching entry-level college life science courses easy and fun for instructor and student. DM500 Brightfield Microscope has plug-and-play capability.
Brand:  Leica Microsystems 1050DM5003
 View more versions of this product
Product Code. 12817975
  • 7380.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
All systems include XY mechanical stage, coarse/fine focus, dust cover, immersion oil and user manual.
  • The ideal tool for making life science university courses easy and fun for both instructor and student
  • The pre-focused, pre-centred condenser and the EZTube™ preset dioptre eyepieces prevent incorrect adjustments and provide more time for hands-on teaching
  • EZStore™ design with integrated handle and cord wrap allows easy carrying and lifting, and protection against microscope component damage
  • Features LED illumination, eliminating the nuisance and expense of replacing bulbs
  • The optical system uses the same platform as Leica research grade microscopes
  • 'Rackless' stage system means there is no rack and pinion mechanism protruding from the side of the stage - this prevents minor scratches or grazes to the skin, or the tearing of gloves
Specifications
Specifications
Prefocused, precentered Abbe condenser
LED
DM5003
Brightfield Microscope
220/240 V 50/60 Hz
Binocular, High Eyepoint
100X
Brightfield, Phase Contrast, Infinity Optics
219 x 282 x 428 mm (standard configuration)