Learn More
Leica Microsystems DM750 Brightfield Microscope
Ideal microscope for the versatile needs of advanced college life science courses and professional training in medical, veterinary and dental schools. Leica Microsystems™ DM750 Brightfield Microscope includes student- and teacher-friendly features.
Brand:  Leica Microsystems 1050DM7501
 View more versions of this product
Product Code. 12897985
  • 6610.00 € / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
All systems include XY mechanical stage, coarse/fine focus, dust cover, immersion oil, and user manual.
  • The ideal choice for advanced college and university life science courses and professional training in medical, veterinary and dental schools
  • EZStore™ design with integrated handle and cord wrap allows easy carrying, easy lifting and protection against microscope component damage
  • The DM750 range feature LED illumination, eliminating the nuisance and expense of replacing bulbs
  • Includes auto-off function that powers down the lighting after 2hr of unuse
  • The optical system uses the same platform as Leica research grade microscopes
  • Rackless stage system means there is no rack and pinion mechanism protruding from the side of the stage - preventing minor scratches or grazes to the skin or gloves
  • Ultra hard wearing stage surface
  • All systems include XY mechanical stage, course/fine focus, 4 position nosepiece, auto-off, two eyepieces 10x/20 (one fixed, one adjustable) dust cover and immersion oil
Specifications
Specifications
DM750