Learn More
Leica Microsystems DM750 Brightfield Microscope
Ideal microscope for the versatile needs of advanced college life science courses and professional training in medical, veterinary and dental schools. Leica Microsystems™ DM750 Brightfield Microscope includes student- and teacher-friendly features.
6610.00 € - 9520.00 €
Product Code | Brand | Model | Objective Types | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Objective Types | Price | Quantity & Availability | ||||
12807995
|
Leica Microsystems
1050DM7503 |
DM7503 | Brightfield, Hi Plan, Infinity Optics |
7295.00 €
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
12827995
|
Leica Microsystems
1050DM7504 |
DM7504 | Brightfield, Phase Contrast, Hi Plan, Infinity Optics |
9520.00 €
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
||||
12897985
|
Leica Microsystems
1050DM7501 |
DM750 |
6610.00 €
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
Description
- EZStore™ design with integrated handle and cord wrap allows easy carrying, easy lifting and protection against damage.
- EZLite™ provides over 20 years of bright, LED illumination, which saves time and cost.
- Auto-off feature: patented time-delay shutoff saves energy.
- AgTreat™ anti-bacteria surface coating.
- The optical system uses the same platform as Leica research grade microscopes.
- Rackless stage system means there is no rack and pinion mechanism protruding from the side of the stage - preventing minor scratches or grazes to the skin or gloves.
- Ultra hard wearing stage surface.
- All systems include XY mechanical stage, course/fine focus, 4-position nosepiece, auto-off, two eyepieces 10x/20 (one fixed, one adjustable), dust cover and immersion oil