missing translation for 'onlineSavingsMsg'
Learn More

BRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18998-1-AP-150UL

 View more versions of this product

Product Code. 16848814

  • 410.00 € / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This gene encodes a member of the POU-III class of neural transcription factors. The encoded protein is involved in neuronal differentiation and enhances the activation of corticotropin-releasing hormone regulated genes. Overexpression of this protein is associated with an increase in the proliferation of melanoma cells.
Specifications

Specifications

BRN2
Polyclonal
Unconjugated
POU3F2
Brain 2, Brn 2, BRN2, Oct 7, OCT7, Octamer binding protein 7, OTF 7, OTF7, POU class 3 homeobox 2, POU3F2, POUF3
Rabbit
Antigen Affinity Chromatography
RUO
18992, 29588, 5454
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20265, P31360, P56222
Pou3f2
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers