missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | BRN2 |
---|---|
Concentration | 0.36 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16858814
|
Proteintech
18998-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16848814
|
Proteintech
18998-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the POU-III class of neural transcription factors. The encoded protein is involved in neuronal differentiation and enhances the activation of corticotropin-releasing hormone regulated genes. Overexpression of this protein is associated with an increase in the proliferation of melanoma cells.Specifications
BRN2 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P20265, P31360, P56222 | |
18992, 29588, 5454 | |
Peptide | |
Primary | |
-20°C | |
POU3F2 |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Brain 2, Brn 2, BRN2, Oct 7, OCT7, Octamer binding protein 7, OTF 7, OTF7, POU class 3 homeobox 2, POU3F2, POUF3 | |
Pou3f2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |