Learn More
Bromotrichloromethane, 97%, Thermo Scientific Chemicals
39.20 € - 133.67 €
Chemical Identifiers
CAS | 75-62-7 |
---|---|
Molecular Formula | CBrCl3 |
Molecular Weight (g/mol) | 198.265 |
MDL Number | MFCD00000783 |
InChI Key | XNNQFQFUQLJSQT-UHFFFAOYSA-N |
Synonym | methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br |
PubChem CID | 6383 |
IUPAC Name | bromo(trichloro)methane |
SMILES | C(Cl)(Cl)(Cl)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479772
|
Thermo Scientific Alfa Aesar
A10824.22 |
100 g |
39.20 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489772
|
Thermo Scientific Alfa Aesar
A10824.36 |
500 g |
133.67 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the bromination of benzylic positions under sunlamp irradiation. Employed as chain transfer agent for radical polymerization of methacrylates. It can also be employed as brominating reagent of active hydrogen compounds.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Protect against electrostatic charges.
Chemical Identifiers
75-62-7 | |
198.265 | |
XNNQFQFUQLJSQT-UHFFFAOYSA-N | |
6383 | |
C(Cl)(Cl)(Cl)Br |
CBrCl3 | |
MFCD00000783 | |
methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br | |
bromo(trichloro)methane |
Specifications
75-62-7 | |
2.015 | |
None | |
CBrCl3 | |
1.506 | |
UN2810 | |
methane, bromotrichloro, trichlorobromomethane, bromo trichloro methane, carbon bromotrichloride, monobromotrichloromethane, carbon trichlorobromide, trichloromethyl bromide, caswell no. 118, unii-ikj30qxm63, ccl3br | |
XNNQFQFUQLJSQT-UHFFFAOYSA-N | |
bromo(trichloro)methane | |
6383 | |
97% |
-6°C | |
104°C to 105°C | |
Chloroform-like | |
BrCCl3 | |
MFCD00000783 | |
1732543 | |
Not miscible or difficult to mix in water. | |
C(Cl)(Cl)(Cl)Br | |
198.265 | |
198.28 | |
Bromotrichloromethane |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 200-886-0
missing translation for 'rtecsNumber' : PA5400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only