Learn More
Hirschmann™ Class A Amber Glass Volumetric Flasks with Poly Stopper
Allows for precise, safe liquid measurement and handling of light-sensitive materials in the laboratory. Hirschmann Instruments™ Class A Amber Glass Volumetric Flasks with Poly Stopper are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2640171
 View more versions of this product
Product Code. 10215443
  • 72.00 € / Pack of 2
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
Brown
Borosilicate Glass, Duran Glass
±0.06 mL
±0.06 mL
A
Polyethylene
25 mL
110 mm
12/21 NS Standard Ground
Yes