Learn More
Thermo Scientific™ BS3 (bis(sulfosuccinimidyl)suberate)
Crosslink amines to amines with BS3; composed of sulfo-NHS-ester groups separated by an 11.4A, water-soluble spacer arm that is useful for cell-surface protein crosslinking.
Marque: Thermo Scientific™ A39266
192.85 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce BS3 (Sulfo-DSS) is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable and membrane impermeable.
- Reactive groups: sulfo-NHS ester (both ends)
- Reactive towards: amino groups (primary amines)
- Amine-reactive Sulfo-NHS ester reacts rapidly with any primary amine-containing molecule
- Water-soluble; compare with DSS
- Membrane-impermeable, allowing for cell surface labeling
- High-purity, crystalline reagent can be used to create high-purity crosslinked conjugates

Spécification
Amine-Amine | |
572.43 | |
11.4 Å | |
No | |
No-Weigh™, Pierce™ | |
Homobifunctional | |
Medium (10 to 30 Å) | |
10 x 2 mg | |
Standard, Single-use, Premium-grade | |
Crosslinker |
No | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
Sulfo-NHS Ester | |
Solid | |
Water | |
Yes |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only. Not for use in diagnostic procedures.