Learn More
BTZ043, MedChemExpress
MedChemExpress BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.
Brand: Medchem Express HY-13579
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.Specifications
BTZ043 | |
Yellow | |
C17H16F3N3O5S | |
DMSO : 13.3 mg/mL (30.83 mM; Need ultrasonic and warming) | |
431.39 | |
99.75% | |
Solid |
1161233-85-7 | |
Research | |
50 mg | |
O=C1N=C(N(CC2)CCC32OC[C@H](C)O3)SC4=C([N+]([O-])=O)C=C(C(F)(F)F)C=C14 | |
431.39 | |
Research | |
Cancer-programmed cell death |