Learn More
BTZ043, MedChemExpress
MedChemExpress BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.
112.05 € - 837.00 €
Specifications
Chemical Name or Material | BTZ043 |
---|---|
CAS | 1161233-85-7 |
Color | Yellow |
Purity Grade Notes | Research |
Molecular Formula | C17H16F3N3O5S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498437
|
Medchem Express
HY-13579 |
5 mg |
112.05 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16461917
|
Medchem Express
HY-13579 |
50 mg |
557.55 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436236
|
Medchem Express
HY-13579 |
100 mg |
837.00 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16415768
|
Medchem Express
HY-13579 |
10 mg |
174.15 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.Specifications
BTZ043 | |
Yellow | |
C17H16F3N3O5S | |
O=C1N=C(N(CC2)CCC32OC[C@H](C)O3)SC4=C([N+]([O-])=O)C=C(C(F)(F)F)C=C14 | |
431.39 | |
Research | |
Cancer-programmed cell death |
1161233-85-7 | |
Research | |
DMSO : 13.3 mg/mL (30.83 mM; Need ultrasonic and warming) | |
431.39 | |
99.75% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.