missing translation for 'onlineSavingsMsg'
Learn More

Notrax Bubble Sof-Tred™ Dyna-Shield™ Layer with Unique Bubble Design

Notrax Bubble Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.

Brand:  Notrax 417C0024BY

 View more versions of this product

Product Code. 17782098

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lengthwise corrugated rib pattern for sure footing
  • Exceptional worker comfort due to 127 mm thick specially formulated compound
  • Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
  • Available with yellow borders along sides comply with OSHA code 1910-144
  • All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications

Specifications

Black/Yellow
1830 cm
Industrial and Commercial Matting
60 ft.
Microcellular Vinyl
60 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers