missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 258600010

51.40 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.


 View more versions of this product

Product Code. 10359240

  • 54.10 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Buffer solution pH 10
1332-57-6, 144-55-8, 7732-18-5
Liquid
Plastic bottle
Colorless
9.98 to 10.02 (20°C)
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer solution pH 10, Carbonate buffer

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only