missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 7, Phosphate buffer, Thermo Scientific Chemicals
25.00 € - 73.00 €
Specifications
Chemical Name or Material | Buffer solution pH 7 |
---|---|
CAS | 7558-79-4, 7732-18-5 |
Molecular Formula | H2KO4P |
MDL Number | MFCD03097944 |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10448273
|
Thermo Scientific Acros
258591000 |
100 mL | Plastic bottle |
25.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10317531
|
Thermo Scientific Acros
258595000 |
500 mL | Plastic bottle |
35.30 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10114680
|
Thermo Scientific Acros
258590010 |
1 L | Plastic bottle |
52.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11906971
|
Thermo Scientific Acros
258590025 |
2.5 L | Plastic bottle |
73.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH bufferSpecifications
Buffer solution pH 7 | |
H2KO4P | |
Plastic bottle | |
14mmHg at 20°C |
7558-79-4, 7732-18-5 | |
MFCD03097944 | |
14, 7659 | |
136.09 |
RUO – Research Use Only