missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 8.00, Reagecon™
Certified by an ISO 17025 accredited test method and NIST traceable. Reagecon™ Buffer Solution, pH 8.00 is available in solutions tested at 20°C or 25°C to an accuracy level of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, even after opening the bottle.
38.20 € - 49.50 €
Specifications
Melting Point | 0°C |
---|---|
Boiling Point | 100°C |
Color | Colorless |
Physical Form | Liquid |
Solution Type | pH Buffer |
Traceability to NIST | Yes |
Solubility Information | Fully miscible in water |
Product Code | Brand | Quantity | pH | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | pH | Price | Quantity & Availability | ||||
11482014
|
Reagecon
10805 |
500 mL | 8 |
45.90 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11822172
|
Reagecon
1080525 |
500 mL | 8 |
43.50 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11420469
|
Reagecon
1080 |
1 L | 8 |
49.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12719166
|
Reagecon
108025 |
1 L | 8 |
38.20 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
0°C | |
Colorless | |
pH Buffer | |
Fully miscible in water |
100°C | |
Liquid | |
Yes |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- BUFFER SOLUTION pH 8.00 ± 0.01 @ 20°C
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Boric Acid, DisodiumSalt