missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid

Brand:  Fisher Chemical J/2840/17

70.64 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 10685682

  • 88.30 € / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7778-77-0
6
2.5 L
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
516951
Colorless
Liquid
H2KO4P
HDPE plastic bottle
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer colour coded concentrated solution pH 10 (borate) Blue

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.