missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid

39.20 € - 88.30 €

Specifications

CAS 7778-77-0
Color Colorless
pH 6
Physical Form Liquid
Packaging HDPE plastic bottle
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10690404
View Documents
Fisher Chemical
J/2840/08
500 mL
39.20 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10000210
missing translation for 'viewDocuments'
Fisher Chemical
J/2840/15
1 L
51.40 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10685682
missing translation for 'viewDocuments'
Fisher Chemical
J/2840/17
2.5 L
88.30 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid

Specifications

Specifications

7778-77-0
6
HDPE plastic bottle
Colorless
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer colour coded concentrated solution pH 10 (borate) Blue

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.