Learn More
Macherey-Nagel™ Buffers for NucleoSpin™ Plasmid Kits
Additional supply of buffer solutions (Resuspension Buffer A1, Lysis Buffer A2, Neutralization Buffer A3, Wash Buffer AW, Wash Buffer A4 Concentrate, or Elution Buffer AE) for use in preparing high-purity plasmid DNA using a variety of NucleoSpin™ Plasmid kits.
Brand:  Macherey-Nagel™ 740914.3
 View more versions of this product
Product Code. 15899127
  • 104.00 € / 600mL
Explore more special offers
Description
Description
For use with the following NucleoSpin™ Plasmid DNA purification kits:
  • NucleoSpin™ Plasmid (Lid/NoLid)
  • NucleoSpin™ Plasmid QuickPure
  • NucleoSpin™ Plasmid EasyPure
  • NucleoSpin™ 8/96 Plasmid (Full/Core Kit)
  • NucleoSpin™ Plasmid Transfection-Grade
  • NucleoSpin™ 96 Plasmid Transfection-Grade (Full/Core Kit)
Specifications
Specifications
Colorless
NucleoSpin™ Plasmid kits
600 mL
Room Temperature (18°C to 25°C)
Preparation high-purity plasmid DNA
4X (Yields 3000 mL working solution)
7.0 to 8.0
Odorless
Liquid
Wash Buffer A4 (Concentrate)