Learn More
Macherey-Nagel™ Buffers for NucleoSpin™ Plasmid Kits
Additional supply of buffer solutions (Resuspension Buffer A1, Lysis Buffer A2, Neutralization Buffer A3, Wash Buffer AW, Wash Buffer A4 Concentrate, or Elution Buffer AE) for use in preparing high-purity plasmid DNA using a variety of NucleoSpin™ Plasmid kits.
25.65 € - 482.00 €
Specifications
Color | Colorless |
---|---|
For Use With (Equipment) | NucleoSpin™ Plasmid kits |
Content And Storage | Room Temperature (18°C to 25°C) |
Form | Liquid |
For Use With (Application) | Preparation high-purity plasmid DNA |
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
15819137
|
Macherey-Nagel™
740917.3 |
Elution Buffer AE | 3000 mL |
217.00 €
3000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15879127
|
Macherey-Nagel™
740912.3 |
Lysis Buffer A2 | 3000 mL |
261.00 €
3000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15889127
|
Macherey-Nagel™
740913.3 |
Neutralization Buffer A3 | 3000 mL |
482.00 €
3000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15859127
|
Macherey-Nagel™
740911.3 |
Resuspension Buffer A1 | 3000 mL |
261.00 €
3000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15869127
|
Macherey-Nagel™
740911.75 |
Resuspension Buffer A1 | 75 mL |
25.65 €
75mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899127
|
Macherey-Nagel™
740914.3 |
Wash Buffer A4 (Concentrate) | 600 mL |
104.00 €
600mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15849147
|
Macherey-Nagel™
740995 |
Wash Buffer AQ Concentrate | 25 mL |
41.70 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15809137
|
Macherey-Nagel™
740916.3 |
Wash Buffer AW | 3000 mL |
482.00 €
3000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For use with the following NucleoSpin™ Plasmid DNA purification kits:
- NucleoSpin™ Plasmid (Lid/NoLid)
- NucleoSpin™ Plasmid QuickPure
- NucleoSpin™ Plasmid EasyPure
- NucleoSpin™ 8/96 Plasmid (Full/Core Kit)
- NucleoSpin™ Plasmid Transfection-Grade
- NucleoSpin™ 96 Plasmid Transfection-Grade (Full/Core Kit)
Specifications
Colorless | |
Room Temperature (18°C to 25°C) | |
Preparation high-purity plasmid DNA |
NucleoSpin™ Plasmid kits | |
Liquid |
Safety and Handling
missing translation for 'shelfLife' : At least 1 year
Your input is important to us. Please complete this form to provide feedback related to the content on this product.