missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco Bulk Tray Dryer
FreeZone Bulk Tray Dryers lyophilize moderate to large bulk sample loads or small batches of vials or serum bottles. FreeZone Console Freeze Dryer and vacuum pump are required.
Brand: Labconco 780601190
This item is not returnable.
View return policy
Description
- Lyophilize moderate to large bulk sample loads or numerous small batches of vials or serum bottles.
- Bulk Tray Dryers have three large, 12.7 x 16.6 in. shelves that heat up to +60°C.
- Lyo-Works™ OS lets the user set up to 30 programs, with up to 16 steps each; record, chart and store data, and receive email alerts.
- The tray dryer connects to a FreeZone Console Freeze Dryer that provides a real time display of the tray dryer’s parameters on a large capacitive touchscreen.
- For use with FreeZone Console Freeze Dryer manufactured after July 2017.
Specifications
Bulk Tray Dryer | |
60°C | |
6-Port Manifold, Bulk Tray Dryer | |
230 V |
3 | |
Stainless Steel | |
All except North America |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Labconco-freezone_brochure.pdf
File size: 15.8 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Labconco Bulk Tray Dryer > Includes: 6-Port Manifold, Bulk Tray Dryer
Spot an opportunity for improvement?Share a Content Correction