Learn More
Juchheim Laborgeraete™ Bunsen Burner
A high level of stability is achieved due to the high weight and low center of gravity of the burners.
Brand:  Juchheim Laborgeraete™ 01150 DP
 View more versions of this product
Product Code. 10432752
  • 33.70 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Choice of Propane gas or Natural gas
  • According to DIN 30665
  • With DVGW certification
  • Heating capacity approx. 2198 kJ/h
  • With air control screen
  • Needle valve option
  • Particularly good stand due to heavy, high-quality materials
Specifications
Specifications
Bunsen Burner
Propane
Brass
Air Regulator