Learn More
Juchheim Laborgeraete™ Bunsen Burner
A high level of stability is achieved due to the high weight and low center of gravity of the burners.
33.70 € - 45.40 €
Product Code | Brand | Gas Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Gas Type | Includes | Price | Quantity & Availability | ||||
10432752
|
Juchheim Laborgeraete™
01150 DP |
Propane | Air Regulator |
33.70 €
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
||||
10482552
|
Juchheim Laborgeraete™
01160 DE |
Natural Gas | Needle Valve |
45.40 €
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
||||
Description
- Choice of Propane gas or Natural gas
- According to DIN 30665
- With DVGW certification
- Heating capacity approx. 2198 kJ/h
- With air control screen
- Needle valve option
- Particularly good stand due to heavy, high-quality materials
Specifications
Bunsen Burner | |
Brass |