Learn More
Juchheim Laborgeraete™ Bunsen Burner
A high level of stability is achieved due to the high weight and low center of gravity of the burners.
Brand: Juchheim Laborgeraete™ 01160 DE
This item is not returnable.
View return policy
Description
- Choice of Propane gas or Natural gas
- According to DIN 30665
- With DVGW certification
- Heating capacity approx. 2198 kJ/h
- With air control screen
- Needle valve option
- Particularly good stand due to heavy, high-quality materials
Specifications
Bunsen Burner | |
Natural Gas | |
Brass | |
Needle Valve |