missing translation for 'onlineSavingsMsg'
Learn More

RL Enterprises™ Bunsen Burner Tube

Brand:  RL Enterprises™ 0311

Product Code. 12940896

  • 87.70 € / Pack of 10
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Specifications

Specifications

Tube
13 mm
10 Pk.
13 mm
Byc-395-N and Byd-410-M
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RL Enterprises™ Bunsen Burner Tube > Size: 13mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.