missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Seals, Tapes, and Foils

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.

Brand:  Thermo Scientific™ AB-0718

79.92 EUR valid until 2025-05-31
Use promo code "25075" to get your promotional price.


 View more versions of this product

Product Code. 10642674

  • 144.00 € / Pack of 50
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

Adhesive Plate Seals provide an effective seal. It features an easy-removal non-adhesive tab and can withstand temperatures between -40°C to 80°C.

Highlights

  • Non-adhesive tab for easy removal of seal
  • Temperature range: -40°C to 80°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Gas Permeable Adhesive Seal
Non-sterile
96
50
General
Abgene
No
-20°C to +80°C
All Microplates
Yes
Life Science Reagent Resistant
Polypropylene
Not Autoclavable
50
Ventilating and storing cell cultures
Adhesive Seal
Acrylic
No
All well shapes
General
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils > -20degreeC to 80degreeC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.