missing translation for 'onlineSavingsMsg'
Learn More

Techsit™ C-Tech ESD Cleanroom Low Chair

Cleanroom ESD anti-static vinyl low chair is an ergonomically designed seat with a back.

Brand:  Techsit™ TS8920AA

Encompass_Preferred

 View more versions of this product

Product Code. 16553944

  • 632.74 € / Each
Estimated Shipment: 05-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ESD Vinyl low chair on casters
  • Black ESD/Anti-static Vinyl that is easy to clean
  • Fully adjustable seat height, back height, and back rack
  • Ergonomically designed and contoured seat and back
  • Cleanroom Class 7 suitable
  • BS EN61340-5-1 Conductivity 104 - 106 Ω
Specifications

Specifications

Cleanroom Seating
Yes
Not Autoclavable
ESD Vinyl
Gas Lift
Desk Height
5 Year
Casters
Yes
Yes
Yes
Vinyl
Black
Adjustable Arms
Ergonomic
Steel
150 kg
Vinyl
16 kg
490 mm
Yes
No
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers