Learn More
Cacodylic acid sodium salt trihydrate 98+%, Thermo Scientific Chemicals
67.00 € - 744.00 €
Chemical Identifiers
CAS | 6131-99-3 |
---|---|
Molecular Formula | C2H12AsNaO5 |
Molecular Weight (g/mol) | 214.024 |
MDL Number | MFCD00149079 |
InChI Key | RLGWPHBPRCROJO-UHFFFAOYSA-M |
Synonym | sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate |
PubChem CID | 23679059 |
IUPAC Name | sodium;dimethylarsinate;trihydrate |
SMILES | C[As](=O)(C)[O-].O.O.O.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473647
|
Thermo Scientific Alfa Aesar
A18139.14 |
25 g |
67.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483647
|
Thermo Scientific Alfa Aesar
A18139.22 |
100 g |
201.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493647
|
Thermo Scientific Alfa Aesar
A18139.36 |
500 g |
744.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
Solubility
Soluble in water and alcohol. Insoluble in diethyl ether.
Notes
Store in cool place. Hygroscopic and moisture sensitive. Incompatible withstrong oxidizing agents and strong bases.
Chemical Identifiers
6131-99-3 | |
214.024 | |
RLGWPHBPRCROJO-UHFFFAOYSA-M | |
23679059 | |
C[As](=O)(C)[O-].O.O.O.[Na+] |
C2H12AsNaO5 | |
MFCD00149079 | |
sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate | |
sodium;dimethylarsinate;trihydrate |
Specifications
6131-99-3 | |
(CH3)2AsO2Na·3H2O | |
UN1688 | |
14,8595 | |
Soluble in water and alcohol. Insoluble in diethyl ether. | |
C[As](=O)(C)[O-].O.O.O.[Na+] | |
214.024 | |
214.03 (159.98 Anhydrous) | |
Cacodylic acid sodium salt trihydrate |
C2H12AsNaO5 | |
MFCD00149079 | |
3702348 | |
sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate | |
RLGWPHBPRCROJO-UHFFFAOYSA-M | |
sodium;dimethylarsinate;trihydrate | |
23679059 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H301+H331-H350-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SODIUM CACODYLATE
missing translation for 'einecsNumber' : 204-708-2
missing translation for 'rtecsNumber' : CH7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Carcinogenicity Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
RUO – Research Use Only