Learn More
Cadmium acetate dihydrate, 99.999% (metals basis), Thermo Scientific Chemicals
183.00 € - 1993.00 €
Chemical Identifiers
CAS | 5743-04-4 |
---|---|
Molecular Formula | C4H10CdO6 |
Molecular Weight (g/mol) | 266.53 |
MDL Number | MFCD00012449 |
InChI Key | AUIZLSZEDUYGDE-UHFFFAOYSA-L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351938
|
Thermo Scientific Alfa Aesar
011865.14 |
25 g | |||||||
11371938
|
Thermo Scientific Alfa Aesar
011865.22 |
100 g | |||||||
11361938
|
Thermo Scientific Alfa Aesar
011865.36 |
500 g |
1993.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in dyestuff intermediates, ceramics and producing iridescent effects on porcelains and pottery. It acts as a reagent for determination of S, Se, and Te. It is also used in cadmium electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5743-04-4 | |
266.53 | |
AUIZLSZEDUYGDE-UHFFFAOYSA-L |
C4H10CdO6 | |
MFCD00012449 |
Specifications
5743-04-4 | |
Powder | |
C4H10CdO6 | |
Soluble in water,ethanol and methanol. | |
266.53 | |
Odorless | |
2.01 |
White | |
99.999% (Metals basis) | |
MFCD00012449 | |
AUIZLSZEDUYGDE-UHFFFAOYSA-L | |
266.54 (230.50 Anhydrous) | |
Hygroscopic | |
Cadmium acetate dihydrate |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H335-H350-H373
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 208-853-2
missing translation for 'rtecsNumber' : AF7505000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only