missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromate hydrate, 98%, Thermo Scientific™

29.00 € - 107.00 €

Chemical Identifiers

CAS 14518-94-6
Molecular Formula Br2CdO6
Molecular Weight (g/mol) 368.22
MDL Number MFCD04971194
InChI Key VECQEXBULIKSDM-UHFFFAOYSA-L
Synonym cadmium bromate, cadmium dibromate, cadmium 2+ dibromate, 2bro3.cd, acmc-1c8r8, bromic acid, cadmiumsalt 8ci,9ci
PubChem CID 14598111
SMILES [Cd++].[O-][Br](=O)=O.[O-][Br](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11325149
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17964.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11335149
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17964.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14518-94-6
368.22
VECQEXBULIKSDM-UHFFFAOYSA-L
14598111
Br2CdO6
MFCD04971194
cadmium bromate, cadmium dibromate, cadmium 2+ dibromate, 2bro3.cd, acmc-1c8r8, bromic acid, cadmiumsalt 8ci,9ci
[Cd++].[O-][Br](=O)=O.[O-][Br](=O)=O

Specifications

14518-94-6
Cd(BrO3)2·xH2O
UN1450
VECQEXBULIKSDM-UHFFFAOYSA-L
cadmium(2+) dibromate
14598111
98%
Br2CdO6
MFCD04971194
cadmium bromate, cadmium dibromate, cadmium 2+ dibromate, 2bro3.cd, acmc-1c8r8, bromic acid, cadmiumsalt 8ci,9ci
[Cd++].[O-][Br](=O)=O.[O-][Br](=O)=O
368.22
368.21 (Anhydrous)
Cadmium bromate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H302-H312-H332
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P210-P220-P221-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P370+P378q-P501c

H272-H302+H312+H332-H350

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BROMATES, INORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications