Learn More
Cadmium bromide tetrahydrate, Reagent Grade, Thermo Scientific Chemicals
46.00 € - 150.00 €
Chemical Identifiers
CAS | 13464-92-1 |
---|---|
Molecular Formula | Br2CdH8O4 |
Molecular Weight (g/mol) | 344.282 |
MDL Number | MFCD00150204 |
InChI Key | PANYFCXNZSEXSU-UHFFFAOYSA-L |
Synonym | cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent |
PubChem CID | 3084147 |
IUPAC Name | cadmium(2+);dibromide;tetrahydrate |
SMILES | O.O.O.O.[Br-].[Br-].[Cd+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343118
|
Thermo Scientific Alfa Aesar
032958.14 |
25 g |
46.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333118
|
Thermo Scientific Alfa Aesar
032958.22 |
100 g |
150.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13464-92-1 | |
344.282 | |
PANYFCXNZSEXSU-UHFFFAOYSA-L | |
3084147 | |
O.O.O.O.[Br-].[Br-].[Cd+2] |
Br2CdH8O4 | |
MFCD00150204 | |
cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent | |
cadmium(2+);dibromide;tetrahydrate |
Specifications
13464-92-1 | |
Br2CdH8O4 | |
UN2570 | |
Soluble in water. | |
O.O.O.O.[Br-].[Br-].[Cd+2] | |
344.282 | |
344.28 (272.22 Anhydrous) | |
Hygroscopic |
Powder | |
MFCD00150204 | |
cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent | |
PANYFCXNZSEXSU-UHFFFAOYSA-L | |
cadmium(2+);dibromide;tetrahydrate | |
3084147 | |
Reagent | |
Cadmium bromide tetrahydrate |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H350
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
missing translation for 'einecsNumber' : 232-165-1
missing translation for 'rtecsNumber' : EU9935000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only