Learn More
Cadmium sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
Used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum.
167.00 € - 736.00 €
Chemical Identifiers
CAS | 1306-23-6 |
---|---|
Molecular Formula | CdS |
Molecular Weight (g/mol) | 144.47 |
MDL Number | MFCD00010922 |
InChI Key | FRLJSGOEGLARCA-UHFFFAOYSA-N |
Synonym | cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow |
PubChem CID | 14783 |
ChEBI | CHEBI:50833 |
IUPAC Name | sulfanylidenecadmium |
SMILES | [S--].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15452267
|
Thermo Scientific Alfa Aesar
045455.18 |
50 g |
167.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15462267
|
Thermo Scientific Alfa Aesar
045455.30 |
250 g |
736.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadmium sulfide is used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum. It is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-23-6 | |
144.47 | |
FRLJSGOEGLARCA-UHFFFAOYSA-N | |
14783 | |
sulfanylidenecadmium |
CdS | |
MFCD00010922 | |
cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
CHEBI:50833 | |
[S--].[Cd++] |
Specifications
980°C (sublimation) | |
Pieces | |
MFCD00010922 | |
cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
FRLJSGOEGLARCA-UHFFFAOYSA-N | |
sulfanylidenecadmium | |
14783 | |
144.46 | |
Plastic bottle | |
4.82 |
1306-23-6 | |
CdS | |
UN3077 | |
Insoluble in water. | |
[S--].[Cd++] | |
144.47 | |
CHEBI:50833 | |
99.999% | |
4.82 | |
Cadmium sulfide |
Safety and Handling
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H341-H350-H361fd-H372
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 215-147-8
missing translation for 'rtecsNumber' : EV3150000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only