missing translation for 'onlineSavingsMsg'
Learn More
Calcium for AAS 1g/L HCl matrix, Fisher Chemical™ Product Code.: 12913135

Calcium for AAS 1g/L HCl matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12913135
100 mL
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
This item is not returnable. View return policy

Product Code. 12913135

Brand: Fisher Chemical B 009 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Bottle
Quantity 100 mL
Physical Form Liquid
Concentration 1 g/L ±0.002 g/L
Type Flame Single-element Standard Solution
Solvent or Matrix 2% HCl
Chemical Name or Material Calcium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1047038 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberB 009 100
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium for AAS 1g/L HCl matrix, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.