missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Acetate Monohydrate, ACS Reagent, ≥99%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Color | White |
Quantity | 100 g |
Linear Formula | Ca(CH3CO2)2 · H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 176.18g/mol |
Percent Purity | ≥99% |
Packaging | Poly Bottle |
Appearance | Powder |
Chemical Name or Material | Calcium acetate monohydrate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium Acetate Monohydrate, ACS Reagent, ≥99%, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction