Learn More
Calcium fluoride, 97%, Thermo Scientific Chemicals
43.30 € - 394.00 €
Chemical Identifiers
CAS | 7789-75-5 |
---|---|
Molecular Formula | CaF2 |
Molecular Weight (g/mol) | 78.08 |
MDL Number | MFCD00010907 |
InChI Key | WUKWITHWXAAZEY-UHFFFAOYSA-L |
Synonym | calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade |
PubChem CID | 84512 |
ChEBI | CHEBI:35437 |
IUPAC Name | calcium;difluoride |
SMILES | [F-].[F-].[Ca++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429472
|
Thermo Scientific Alfa Aesar
A10176.30 |
250 g |
43.30 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419472
|
Thermo Scientific Alfa Aesar
A10176.0B |
1000 g |
118.00 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439472
|
Thermo Scientific Alfa Aesar
A10176.0I |
5000 g |
394.00 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCalcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.
Solubility
Soluble in ammonium salts. Soluble in water (slightly).
Notes
Store at room temperature. Separated from mineral acids. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
Chemical Identifiers
7789-75-5 | |
78.08 | |
WUKWITHWXAAZEY-UHFFFAOYSA-L | |
84512 | |
calcium;difluoride |
CaF2 | |
MFCD00010907 | |
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade | |
CHEBI:35437 | |
[F-].[F-].[Ca++] |
Specifications
∼2500°C | |
7789-75-5 | |
MFCD00010907 | |
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade | |
WUKWITHWXAAZEY-UHFFFAOYSA-L | |
calcium;difluoride | |
84512 | |
78.08 | |
Odorless | |
3.18 |
1403°C | |
CaF2 | |
14,1667 | |
Soluble in ammonium salts. Soluble in water (slightly). | |
[F-].[F-].[Ca++] | |
78.08 | |
CHEBI:35437 | |
97% | |
1.434 | |
Calcium fluoride, Precipitated |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 232-188-7
missing translation for 'rtecsNumber' : EW1760000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only