missing translation for 'onlineSavingsMsg'
Learn More

Calcium shot, 99.5% (metals basis), Thermo Scientific™

22.00 €

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351178
View Documents
Thermo Scientific Alfa Aesar
010127.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7440-70-2
40.08
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
calcium
Ca
MFCD00010897 MFCD00085314
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
CHEBI:29320
[Ca]

Specifications

Silver-White
Silvery-white shot
Ca
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
[Ca]
40.08
CHEBI:29320
0 hPa
Calcium
7440-70-2
99.5% (Metals basis)
MFCD00010897 MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
calcium
5460341
Odorless
Shot
Videos
Safety and Handling

Safety and Handling

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H261

missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: II

missing translation for 'einecsNumber' : 213-179-5

missing translation for 'rtecsNumber' : EV8040000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Calcium shot

Signal Word
  • Danger

Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H261-In contact with water releases flammable gases.

Precautionary Statement
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.