Learn More
Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis), Thermo Scientific Chemicals
53.27 € - 567.83 €
Chemical Identifiers
CAS | 7778-18-9 |
---|---|
Molecular Formula | CaO4S |
Molecular Weight (g/mol) | 136.13 |
MDL Number | MFCD00010912 |
InChI Key | OSGAYBCDTDRGGQ-UHFFFAOYSA-L |
Synonym | calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous |
PubChem CID | 24497 |
ChEBI | CHEBI:31346 |
IUPAC Name | calcium;sulfate |
SMILES | [Ca++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11326138
|
Thermo Scientific Alfa Aesar
011116.04 |
2 g |
53.27 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336138
|
Thermo Scientific Alfa Aesar
011116.09 |
10 g |
161.81 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346138
|
Thermo Scientific Alfa Aesar
011116.18 |
50 g |
567.83 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium sulfate, anhydrous is used as adhesives and sealant chemical, Fillers, Fuels and fuel additives, Intermediates, Process regulators, Agricultural Products, Building/Construction Materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7778-18-9 | |
136.13 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
24497 | |
calcium;sulfate |
CaO4S | |
MFCD00010912 | |
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous | |
CHEBI:31346 | |
[Ca++].[O-]S([O-])(=O)=O |
Specifications
1450°C | |
White | |
CaO4S | |
MFCD00010912 | |
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium;sulfate | |
24497 | |
136.14 | |
Puratronic™ | |
Hygroscopic | |
(metals basis) |
7778-18-9 | |
Powder | |
CaSO4 | |
14,1706 | |
Soluble in water,ammonium salts,sodium thiosulfate & glycerin. | |
[Ca++].[O-]S([O-])(=O)=O | |
136.13 | |
CHEBI:31346 | |
99.993% | |
Odorless | |
2.960 g/mL | |
Calcium sulfate, Anhydrous |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
missing translation for 'einecsNumber' : 231-900-3
missing translation for 'rtecsNumber' : WS6920000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only