missing translation for 'onlineSavingsMsg'
Learn More
Calibration Buffer Solutions, Hanna Instruments™ Product Code.: 10356151

Calibration Buffer Solutions, Hanna Instruments™

Product Code. 10356151
20 mL, 4.01
Click to view available options
Quantity:
20 mL
pH:
4.01
10.01
This item is not returnable. View return policy

Product Code. 10356151

Brand: Hanna Instruments HI70004P

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Premium quality calibration buffer solution with reported values that are accurate to +/- 0.01 pH @ 25°C.

  • Premium quality pH 4.01 calibration buffer solution
  • Color coded label for easy pH buffer value identification
  • Temperature chart of the actual pH value at various temperatures printed on each sachet
  • pH 4.01 calibration buffer produced in accordance with ISO 3696/BS3978 standards
  • Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
  • Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
  • Complete with a pH electrode, temperature probe, calibration buffers, reference refill electrolyte, cleaning solutions, and a probe holder

Specifications

Color Colorless
pH 4.01
Physical Form Liquid
Quantity 20 mL
Solution Type Buffer
Formulation Aqueous Buffer Solution
Odor Odorless
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calibration Buffer Solutions, Hanna Instruments™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.