Learn More
Mettler Toledo™ Buffer Solutions: Standard pH

Storage and refill solutions for pH electrodes
Brand: Mettler Toledo™ 51302069
68.88 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Completely miscible
- Comes with Quality inspection certificate
- NIST traceable

Specifications
∼100°C | |
Buffer Solution | |
Quality inspection certificate | |
4 |
Clear, Red | |
pH Buffer | |
30 x 20 mL Sachets | |
Completely Miscible |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.