Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory
Brand: Hirschmann™ 9212380
Description
- Capillary tubes
- One end closed
- Available in a wide range of diameters/lengths
Specifications
Melting Point Determination Tube, One End Closed | |
1.55 mm | |
80 mm | |
1 x 1000/Pk. |