missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ S-CLASSIC-90 2-Door Yellow Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas, with an optional door open arrest system.
Brand: Asecos™ 3011600233719
This item is not returnable.
View return policy
Description
- Aseco’s first EN-compliant safety storage cabinet
- Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
- Available in a range of widths and heights
- Versatile fields of application due to various interior equipment
- Convenient handling – door operation with minimal effort
- Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
- Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
- Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- In accordance with EN 14470-1 and TRGS 510
Specifications
EN 14470-1 | |
616 x 1196 x 1968 mm | |
Storage of flammable and hazardous substances | |
3 x Tray shelves 150 kg/33 L each; Insert 135 kg; Sump 55 L | |
Powder-coated | |
Yellow | |
Safety storage cabinet |
3 x 150 kg/33 L sheet steel tray shelves; 1 x 135 kg sheet steel insert; 1 x 55 L sheet steel sump | |
Triple-hinged | |
Door open arrest system | |
2 | |
457.5 kg | |
Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Asecos™ S-CLASSIC-90 2-Door Yellow Safety Storage Cabinets > 3 33 L shelves; 1 135 kg insert; 1 55 L sump
Spot an opportunity for improvement?Share a Content Correction