missing translation for 'onlineSavingsMsg'
Learn More
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals Product Code.: 10742601

ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals

Product Code. 10742601
1 kg, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10742601

Brand: Thermo Scientific Acros 173440010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-44-3
MDL Number MFCD00003267
InChI Key PAPBSGBWRJIAAV-UHFFFAOYSA-N
Synonym 6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptaneShow MoreShow Less
PubChem CID 10401
ChEBI CHEBI:17915
IUPAC Name oxepan-2-one
SMILES C1CCC(=O)OCC1

Specifications

Melting Point -1°C
Color Colorless
Density 1.0300g/mL
Boiling Point 235°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H10O2
Refractive Index 1.4620 to 1.464
Quantity 1 kg
Beilstein 17,II,29
Specific Gravity 1.03
Solubility Information Solubility in water: >1000mg/l (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether
Molecular Weight (g/mol) 114.14
Formula Weight 114.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material ε-Caprolactone monomer
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462456 Certificate TypeCertificate of Analysis Date01/05/2024 Catalog Number173440025,173442500,173440010
Lot NumberA0460745 Certificate TypeCertificate of Analysis Date04/03/2024 Catalog Number173440025,173442500,173440010
Lot NumberA0457489 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number173440025,173442500,173440100,173440010
Lot NumberA0453526 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number173440025,173442500,173440100,173440010
Lot NumberA0449672 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number173440025,173442500,173440100,173440010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.