missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Caps for Nunclon™ Delta Flasks
BIOPHARMA

Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close, filter—to meet application needs.

67.31 € - 114.00 €

Specifications

Closure Color Blue
Material HDPE
No. per Case 100
No. per Pack 1
Sterility Sterile
View More Specs

Products 6
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10357351
View Documents
Thermo Scientific™
158892
25 cm² EasY Flask
103.00 €
Pack of 100
Estimated Shipment: 07-08-2025
to see stock.
Add to basket
 
10574751
View Documents
Thermo Scientific™
157527
75 cm² EasY Flask
67.55 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10654492
BIOPHARMA
View Documents
Thermo Scientific™
133001
175 cm² EasY Flask
99.55 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10154271
View Documents
Thermo Scientific™
151152
25 cm² Nunclon Delta Flask N/A N/A N/A
10203981
View Documents
Thermo Scientific™
144458
80 cm² Nunclon Delta Flask
112.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10090611
View Documents
Thermo Scientific™
147104
175/185/500 cm² Nunclon Delta Flask
114.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close (solid) and filter—to meet your application needs.

Vent/close Caps
  • EasY Flask vent/close caps feature a “Y” mark that allows visual verification
  • Vent position easily distinguished even what flasks are stacked in incubators
Filter Caps
  • Filter membrane provides effective barrier against microorganisms
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
Certifications:
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
Ordering Information:
  • Case of 100
Warranty: 90 days
Specifications

Specifications

Blue
100
Sterile
HDPE
1
Nunclon Delta
Videos
SDS
Documents

Documents

Product Certifications

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Caps for Nunclon™ Delta Flasks

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.