Learn More
Erlab™ Captair™ Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Work Surfaces are for use with Captair PCR work stations.
Brand:  Erlab™ I0120020002
 View more versions of this product
Product Code. 12167710
  • 1478.70 € / Each
Explore more special offers
Description
Description
  • High chemical and mechanical resistance
  • Rounded corners to facilitate cleaning operations
  • Built in spill tray
Specifications
Specifications
For Captair™ Flex™ Model XLS 714 fume hood
No. 1 Sealed Tempered Glass Retention Tray, Embedded Metal, Sealed Frame
Work surface No. 1
Glass