Learn More
Erlab™ Captair™ Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Work Surfaces are for use with Captair PCR work stations.
667.80 € - 1881.50 €
Products 9
Product Code Brand Includes For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Includes For Use With (Equipment) Price Quantity & Availability  
12010502
View Documents
Erlab™
I0121020002
Sealed Tempered Glass Retention Tray For Captair™ Flex™ Model M481 fume hood
832.10 €
Each
12167710
View Documents
Erlab™
I0120020002
No. 1 Sealed Tempered Glass Retention Tray, Embedded Metal, Sealed Frame For Captair™ Flex™ Model XLS 714 fume hood
1478.70 €
Each
12361793
View Documents
Erlab™
I0115020002
No. 1 Sealed Tempered Glass Retention Tray, Embedded Metal For Captair™ Flex™ Model Midcap M321 and flow 321 fume hood
667.80 €
Each
12333713
View Documents
Erlab™
I0116030001
No. 1 Sealed Tempered Glass Retention Tray, Sealed Frame For Captair™ Flex™ Model M391 and flow 391 fume hood
1102.40 €
Each
12030472
View Documents
Erlab™
I0116020002
No. 1 Sealed Tempered Glass Retention Tray, Sealed Frame For Captair™ Flex™ Model M391 fume hood
731.40 €
Each
12090492
View Documents
Erlab™
I0118020002
No. 1 Sealed Tempered Glass Retention Tray, Embedded Metal, Sealed Frame For Captair™ Flex™ Model XLS 483 fume hood
1028.20 €
Each
12878733
View Documents
Erlab™
I0117030001
Built-in Retention Tray For Captair™ Flex™ Model XLS392 fume hood
1219.00 €
Each
12808743
View Documents
Erlab™
I0120030001
Sealed Frame For Model XLS71 Laboratory Furniture
1881.50 €
Each
12898733
View Documents
Erlab™
I0119030001
Sealed Frame For Model XLS63 Laboratory Furniture
1595.30 €
Each
Description
Description
  • High chemical and mechanical resistance
  • Rounded corners to facilitate cleaning operations
  • Built in spill tray