Learn More
Erlab™ Captair™ Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Work Surfaces are for use with Captair PCR work stations.
Brand:  Erlab™ I0119030001
 View more versions of this product
Product Code. 12898733
  • 1595.30 € / Each
Explore more special offers
Description
Description
  • High chemical and mechanical resistance
  • Rounded corners to facilitate cleaning operations
  • Built in spill tray
Specifications
Specifications
For Model XLS63 Laboratory Furniture
Table Top
Ideal for Precision Weighing
Sealed Frame
Phenolic Resin